Learn More
Applied Biosystems™ 21 CFR Part 11 Software Module for ViiA™ 7 System
Shop All Applied Biosystems ProductsDescription
This software module supports compliance with 21CFR Part 11 FDA guidelines on electronic records security and prevents unauthorized access to your ViiA7 instruments. When loaded on the instrument, this software will:
• Authenticate each user’s log-in information
• Lock out access and requite re-authentication if user is inactive for a set period of time
• Log any unauthorized attempts to access ViiA7 System software in the Event Log
• Verify each user’s permission to perform predefined Controlled Activities.
• Permits E-signature function for data verification.
Specifications
Specifications
| Content And Storage | One Software CD. |
| For Use With (Equipment) | ViiA™ 7 System |
| Product Line | ViiA |
| Software Type | Instrument Access and Security |
| Product Type | Software Module |
| For Use With (Application) | Real Time PCR (qPCR) |
| Operating System | Windows |
| Quantity | 1 software |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.