missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 21 CFR Part 11 Software Module for QuantStudio™ 12K Flex System

Product Code. 15823384 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15823384 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15823384 Supplier Applied Biosystems™ Supplier No. 4472053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This software module for the QuantStudio™ 12K Flex Real-Time PCR System supports compliance with 21 CFR Part 11 FDA guidelines on electronic records security and prevents unauthorized access to the instrument.

This software module for the QuantStudio™ 12K Flex Real-Time PCR System supports compliance with 21 CFR Part 11 FDA guidelines on electronic records security and prevents unauthorized access to the instrument. When loaded on the system, this software will:

• Authenticate each user’s log-in information
• Lock out access and require re-authentication if user is inactive for a set period of time
• Log any unauthorized attempts to access the system software in the Event Log
• Verify each user’s permission to perform predefined Controlled Activities
• Permits E-signature function for data verification

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) QuantStudio™ 12 K Flex
Product Line QuantStudio
Product Type Software Module
For Use With (Application) Real Time PCR (qPCR)
Operating System Windows 7
License 10 Seat
Quantity 1 kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.