missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand Tank Level Sensor 200L

Brand:  Fisherbrand RAPF20022

Tank Level Sensor 200L

Product Code. 15479659

  • ONLINE EXCLUSIVE PRICE
    412.00 EUR
    332.54 EUR / Each
    Save 79.46 EUR
    19% Off

Please to purchase this item. Need a web account? Register with us today!


Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.