Learn More
20 cSt Mineral Oil Blank - Trace metals and sulfur reported, VHG™
Description
20 cSt Mineral Oil Blank - Trace metals and sulfur reported
Specifications
Specifications
| Chemical Name or Material | 20 cSt Mineral Oil Blank |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | 20 cSt Mineral Oil Blank |
| Shipping Condition | Room Temperature |
| IUPAC Name | 20 cSt Mineral Oil Blank |
| Solvent or Matrix | 20 cSt Mineral Oil |
| Quantity | 1 gal |