Learn More
Lenz Laborglasintrumente 2-Way Stopcocks with Solid Glass Plug
Product Code. 10020251
Click to view available options
Plug Bore:
2.5 mm (Cap)
Unit Size:
Each
Product Code. 10020251
Brand: Lenz Laborglasintrumente 02461402
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With solid glass plug and screwthread retaining device
  • Two oblique bores
  • Material: borosilicate glass 3.3
  • Leakage rate acc. to DIN 12540-1
Specifications
Plug Bore 2.5 mm (Cap)
Material Borosilicate Glass (3.3)