missing translation for 'onlineSavingsMsg'
Learn More

2-Stearoyl-sn-glycero-3-phosphocholine, TRC

Product Code. 30423193
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423193 10 mg 10mg
30409588 2.5 mg -
30419577 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423193 Supplier Toronto Research Chemicals Supplier No. TRCS68657510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4421-58-3
Molecular Formula C26 H54 N O7 P
Molecular Weight (g/mol) 523.68
Synonym 3,5,8-Trioxa-4-phosphahexacosan-1-aminium, 4-hydroxy-7-(hydroxymethyl)-N,N,N-trimethyl-9-oxo-, inner salt, 4-oxide, (7R)-, 3,5,8-Trioxa-4-phosphahexacosan-1-aminium, 4-hydroxy-7-(hydroxymethyl)-N,N,N-trimethyl-9-oxo-, inner salt, 4-oxide, (R)-, Choline, hydroxide, dihydrogen phosphate, inner salt, 1-ester with 2-monostearin, L- (8CI), (7R)-4-Hydroxy-7-(hydroxymethyl)-N,N,N-trimethyl-9-oxo-3,5,8-trioxa-4-phosphahexacosan-1-aminium inner salt 4-oxide, 2-Stearoyl-sn-glycero-3-phosphocholine
IUPAC Name [(2R)-3-hydroxy-2-octadecanoyloxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCCCC(=O)O[C@H](CO)COP(=O)([O-])OCC[N+](C)(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C26H54NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-26(29)34-25(23-28)24-33-35(30,31)32-22-21-27(2,3)4/h25,28H,5-24H2,1-4H3/t25-/m1/s1
Formula Weight 523.3638
Chemical Name or Material 2-Stearoyl-sn-glycero-3-phosphocholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.