missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2'-OMe-U Phosphoramidite, standard grade, 20-400 finish bottle

Product Code. 17947192 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 17947192

Brand: Thermo Scientific™ 27182510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2'-OMe-U CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) with 2'-O-methyl protection, for use in oligonucleotide synthesis.

2'-OMe-U CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 30-mL amber bottle with a 20-400 finish neck style, which is compatible with most Expedite synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyl-uridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C40H56N7O9P
Base: U
Formula weight: 760.8 g/mol
CAS number: 110764-79-9
HPLC purity: ≥98.0%; Sum of non-primary peaks at 140-152ppm: ≤0.5%
31P NMR purity: ≥98.0%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Expedite Synthesizers
Product Type 2in-OMe-U CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Uridine (U)
Reagent Features 2in-OMe modified
Shipping Condition Room Temperature
Purity 98%
Quantity 1 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type RNA
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.