Learn More
2-O-Desmethyl Cisapride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-O-Desmethyl Cisapride |
| CAS | 102671-04-5 |
| Molecular Formula | C22H27ClFN3O4 |
| Quantity | 25 mg |
| Synonym | cis-4-Amino-5-chloro-N-[1-[3-(4-fluorophenoxy)propyl]-3-methoxy-4-piperidinyl]-2-hydroxy-benzamide, rel-4-Amino-5-chloro-N-[1-[(3R,4S)-3-(4-fluorophenoxy)propyl]-3-methoxy-4-piperidinyl]-2-hydroxybenzamide, 2-O-Desmethyl Acenalin, 2-O-Desmethyl Alimix, 2-O-Desmethyl Cipril, 2-O-Desmethyl Coordinax, 2-O-Desmethyl Prepulsid, 2-O-Desmethyl Propulsid, 2-O-Desmethyl Propulsin |
| InChI Formula | InChI=1S/C22H27ClFN3O4/c1-30-21-13-27(8-2-10-31-15-5-3-14(24)4-6-15)9-7-19(21)26-22(29)16-11-17(23)18(25)12-20(16)28/h3-6,11-12,19,21,28H,2,7-10,13,25H2,1H3,(H,26,29)/t19-,21+/m1/s1 |
| SMILES | CO[C@H]1CN(CC[C@H]1NC(=O)C2=CC(=C(C=C2O)N)Cl)CCCOC3=CC=C(C=C3)F |
| Molecular Weight (g/mol) | 451.92 |
| Formula Weight | 451.1674 |