missing translation for 'onlineSavingsMsg'
Learn More

2’-O-{[(2,2,2,-Trichloroethyl)oxy]carbonyl-7-O-(triethylsilyl) Paclitaxel, TRC

Product Code. 30453444
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453444 100 mg 100mg
30454287 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30453444 Supplier Toronto Research Chemicals Supplier No. TRCT774215100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2’-O-[(2,2,2,-Trichloroethyl)oxy]carbonyl-7-O-(triethylsilyl) Paclitaxel
Analyte or Component Names 2’-O-[(2,2,2,-Trichloroethyl)oxy]carbonyl-7-O-(triethylsilyl) Paclitaxel
Shipping Condition Room Temperature
Molecular Formula C56 H66 Cl3 N O16 Si
Synonym (alphaR,betaS)-beta-(Benzoylamino)-alpha-[[(2,2,2-trichloroethoxy)carbonyl]oxy]-benzenepropanoic Acid (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-Bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4-O-(triethylsilyl)-11-hydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl Ester
InChI Formula InChI=1S/C56H66Cl3NO16Si/c1-10-77(11-2,12-3)76-39-28-40-54(30-69-40,75-34(6)62)45-47(74-49(65)37-26-20-15-21-27-37)55(68)29-38(32(4)41(52(55,7)8)43(71-33(5)61)46(63)53(39,45)9)72-50(66)44(73-51(67)70-31-56(57,58)59)42(35-22-16-13-17-23-35)60-48(64)36-24-18-14-19-25-36/h13-27,38-40,42-45,47,68H,10-12,28-31H2,1-9H3,(H,60,64)/t38-,39-,40+,42-,43+,44+,45-,47-,53+,54-,55+/m0/s1
Solution Type Neat
SMILES CC[Si](CC)(CC)O[C@H]1C[C@H]2OC[C@@]2(OC(=O)C)[C@H]3[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](OC(=O)OCC(Cl)(Cl)Cl)[C@@H](NC(=O)c6ccccc6)c7ccccc7)C(=C([C@@H](OC(=O)C)C(=O)[C@]13C)C5(C)C)C
IUPAC Name [(1S,2S,3R,4S,7R,9S,10S,12R,15S)-4,12-diacetyloxy-15-[(2R,3S)-3-benzamido-3-phenyl-2-(2,2,2-trichloroethoxycarbonyloxy)propanoyl]oxy-1-hydroxy-10,14,17,17-tetramethyl-11-oxo-9-triethylsilyloxy-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] benzoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 1143.565
Quantity 100 mg
Formula Weight 1141.322
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.