Learn More
2-N,N-Dibenzyl Serine Benzyl Ester, TRC
Product Code. 30179820
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179820 1 g 1g
30179777 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30179820 Supplier Toronto Research Chemicals Supplier No. TRCD4186751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Formula Weight 375.183
InChI Formula InChI=1S/C24H25NO3/c26-18-23(24(27)28-19-22-14-8-3-9-15-22)25(16-20-10-4-1-5-11-20)17-21-12-6-2-7-13-21/h1-15,23,26H,16-19H2
IUPAC Name benzyl 2-(dibenzylamino)-3-hydroxypropanoate
Molecular Formula C24 H25 N O3
Molecular Weight (g/mol) 375.46
SMILES OCC(N(Cc1ccccc1)Cc2ccccc2)C(=O)OCc3ccccc3
Synonym N,N-Bis(phenylmethyl)serine Phenylmethyl Ester;
Chemical Name or Material 2-N,N-Dibenzyl Serine Benzyl Ester
Specifications
Quantity 1 g
Shipping Condition Room Temperature