Learn More
2-Methyldecane, TRC
Product Code. 30481246
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481246 1 g 1g
30483143 100 mg 100mg
30469834 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30481246 Supplier Toronto Research Chemicals Supplier No. TRCM0912051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6975-98-0
Molecular Formula C11H24
Molecular Weight (g/mol) 156.31
Synonym Isoundecane, Isopah H, Isopar G
IUPAC Name 2-methyldecane
SMILES CCCCCCCCC(C)C
Specifications
Quantity 1 g
InChI Formula InChI=1S/C11H24/c1-4-5-6-7-8-9-10-11(2)3/h11H,4-10H2,1-3H3
Formula Weight 156.1878
Chemical Name or Material 2-Methyldecane