missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-(trifluoromethyl)pyridine-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Product Code. p-7031870
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15408325

Brand: Thermo Scientific Alfa Aesar H33110.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Participate in Horner-Wadsworth-Emmons reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Participate in Horner-Wadsworth-Emmons reaction.

Solubility
Sparingly Soluble (6.1E-4 g/L) (25°C).

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 545394-83-0
Molecular Formula C8H6F3NO
Molecular Weight (g/mol) 189.14
MDL Number MFCD11035943
InChI Key HHGVNSFXEFHMGQ-UHFFFAOYSA-N
Synonym 2-methyl-6-trifluoromethyl nicotinaldehyde, 2-methyl-6-trifluoromethyl pyridine-3-carbaldehyde, 2-methyl-6-trifluoromethyl pyridine-3-carboxaldehyde, 2-methyl-6-trifluoromethyl-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde, 2-methyl-6-trifluoromethyl
PubChem CID 28875322
IUPAC Name 2-methyl-6-(trifluoromethyl)pyridine-3-carbaldehyde
SMILES CC1=C(C=O)C=CC(=N1)C(F)(F)F

Specifications

Melting Point 45°C to 48°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Sparingly Soluble (6.1E-4g/L) (25°C).
Formula Weight 189.14
Percent Purity 97%
Chemical Name or Material 2-Methyl-6-(trifluoromethyl)pyridine-3-carboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.