missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl chloroformate, tech. 85%

Product Code. p-7028555
Change view
Click to view available options
Quantity:
5 mL
25 mL
250 mL
Unit Size:
250mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455700 5 mL 5mL
11465700 25 mL 25mL
11475700 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455700 Supplier Thermo Scientific Alfa Aesar Supplier No. H25937.AA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 628-12-6 | C4H7ClO3 | 138.547 g/mol

2-methoxyethyl chloroformate was used to produce di-2-dimethoxyethyl hydrazine carboxylate(alternative reagent for the mitsunobu reaction).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-methoxyethyl chloroformate was used to produce di-2-dimethoxyethyl hydrazine carboxylate(alternative reagent for the mitsunobu reaction).

Solubility
Soluble in most organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-12-6
Molecular Formula C4H7ClO3
Molecular Weight (g/mol) 138.547
MDL Number MFCD00058932
InChI Key NDYYWMXJZWHRLZ-UHFFFAOYSA-N
Synonym 2-methoxyethyl chloroformate, chloroformic acid 2-methoxyethyl ester, 2-methoxyethylchloroformate, chloro 2-methoxyethoxy methanone, carbonochloridic acid, 2-methoxyethyl ester, 2-methoxyethylchlorocarbonat, pubchem12774, methoxyethyl chloroformate, acmc-1awq9, 2-methoxy-ethyl chloroformate
PubChem CID 69400
IUPAC Name 2-methoxyethyl carbonochloridate
SMILES COCCOC(=O)Cl

Specifications

Boiling Point 157°C to 158°C
Flash Point 59°C (138°F)
Quantity 5 mL
UN Number UN2742
Formula Weight 138.55
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Methoxyethyl chloroformate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.