missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-5-methoxybenzimidazole, 99%

Product Code. p-7025199
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11458957

Brand: Thermo Scientific Alfa Aesar B20570.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37052-78-1 | C8H8N2OS | 180.23 g/mol

2-Mercapto-5-methoxybenzimidazole is used in the preparation of COX-2 inhibitors. It is also employed in the preparation of compounds such as 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole and 5-methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl-1-oxide)methyl]sulfonyl]-1H-benzimidazole. Further, it acts as a hetercocyclic building block and serves as an intermediate of drug omeprazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercapto-5-methoxybenzimidazole is used in the preparation of COX-2 inhibitors. It is also employed in the preparation of compounds such as 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole and 5-methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl-1-oxide)methyl]sulfonyl]-1H-benzimidazole. Further, it acts as a hetercocyclic building block and serves as an intermediate of drug omeprazole.

Solubility
Soluble in dimethyl sulfoxide. Slightly soluble in methanol. Insoluble in water.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents, acids, bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37052-78-1
Molecular Formula C8H8N2OS
Molecular Weight (g/mol) 180.23
MDL Number MFCD00134581
InChI Key KOFBRZWVWJCLGM-UHFFFAOYSA-N
Synonym 5-methoxy-2-mercaptobenzimidazole, 2-mercapto-5-methoxybenzimidazole, 5-methoxy-1h-benzo d imidazole-2-thiol, 5-methoxy-2-benzimidazolethiol, 5-methoxy-1h-benzimidazole-2-thiol, 5-methoxy-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-methoxy-1h-benzimidazole, 5-methoxy-1h-benzoimidazole-2-thiol, 1,3-dihydro-5-methoxy-2h-benzimidazole-2-thione, 5-methoxybenzimidazole-2-thiol
PubChem CID 665603
SMILES COC1=CC=C2NC(=S)NC2=C1

Specifications

Melting Point 255°C to 259°C
Quantity 5 g
Beilstein 3605123
Solubility Information Soluble in dimethyl sulfoxide. Slightly soluble in methanol. Insoluble in water.
IUPAC Name 5-methoxy-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Formula Weight 180.23
Percent Purity 99%
Chemical Name or Material 2-Mercapto-5-methoxybenzimidazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.