Learn More
2-Iodobenzoyl chloride, 98%
Product Code. 11460054
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460054 50 g 50g
11450054 10 g 10g
11470054 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11460054 Supplier Thermo Scientific Alfa Aesar Supplier No. A13558.18
Request Bulk or Custom Format
CAS: 609-67-6 | C7H4ClIO | 266.462 g/mol
2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodobenzoyl chloride has been used in palladium-catalyzed synthesis of carbomethoxy functional group-induced isoindolin-1-ones, preparation of starting materials required for the synthesis of novel cyclic derivatives of pentavalent iodine, benziodazole oxides.
Solubility
Reacts with water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 609-67-6
Molecular Formula C7H4ClIO
Molecular Weight (g/mol) 266.462
MDL Number MFCD00001040
InChI Key MVIVDSWUOGNODP-UHFFFAOYSA-N
PubChem CID 69112
IUPAC Name 2-iodobenzoyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)I
Specifications
Melting Point 27°C to 32°C
Boiling Point 105°C to 106°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 50 g
UN Number UN3261
Beilstein 2042672
Sensitivity Moisture sensitive
Synonym o-iodobenzoyl chloride, benzoyl chloride, 2-iodo, iodobenzoyl chloride, o-iodobenzoylchloride, 2-iodobezoyl chloride, o-iodo-benzoylchloride, 2-iodobenzoic chloride, 2-iodo-benzoyl chloride, acmc-1atjg, benzoyl chloride,2-iodo
Solubility Information Reacts with water.
Formula Weight 266.47
Percent Purity 98%
Chemical Name or Material 2-Iodobenzoyl chloride