missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-6-methylbenzoic Acid, TRC

Product Code. 30347419
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347419 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347419 Supplier Toronto Research Chemicals Supplier No. TRCB448868200MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Iodo-6-methylbenzoic Acid
CAS 54811-50-6
Molecular Formula C8H7IO2
Quantity 200 mg
InChI Formula InChI=1S/C8H7IO2/c1-5-3-2-4-6(9)7(5)8(10)11/h2-4H,1H3,(H,10,11)
SMILES Cc1cccc(I)c1C(=O)O
IUPAC Name 2-iodo-6-methylbenzoic acid
Molecular Weight (g/mol) 262.04
Formula Weight 261.9491
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.