missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy Trimipramine, TRC

Product Code. 30406128
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406128 5 mg
30401029 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30406128 Supplier Toronto Research Chemicals Supplier No. TRCH9735005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2064-15-5
Molecular Formula C20 H26 N2 O
Molecular Weight (g/mol) 310.43
IUPAC Name 11-[3-(dimethylamino)-2-methylpropyl]-5,6-dihydrobenzo[b][1]benzazepin-3-ol
SMILES CC(CN(C)C)CN1c2ccccc2CCc3cc(O)ccc13

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H26N2O/c1-15(13-21(2)3)14-22-19-7-5-4-6-16(19)8-9-17-12-18(23)10-11-20(17)22/h4-7,10-12,15,23H,8-9,13-14H2,1-3H3
Formula Weight 310.2045
Chemical Name or Material 2-Hydroxy Trimipramine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.