missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy atorvastatin calcium salt, MedChemExpress

Product Code. 16411319
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16411319 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411319 Supplier Medchem Express Supplier No. HY128828

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 2-Hydroxy atorvastatin calcium salt is a hydroxy metabolite of Atorvastatin calcium salt. Atorvastatin is a potent HMG-CoA reductase inhibitor with an IC50 value of 8 nM.

2-Hydroxy atorvastatin calcium salt is a hydroxy metabolite of Atorvastatin calcium salt. Atorvastatin is a potent HMG-CoA reductase inhibitor with an IC50 value of 8 nM.

Specifications

Chemical Name or Material 2-Hydroxy atorvastatin calcium salt
CAS 265989-46-6
Color White
Purity Grade Notes Research
Molecular Formula C33H35FN2O6.1/2Ca
Quantity 1 mg
Solubility Information DMSO : 12.5 mg/mL (21.02 mM; ultrasonic and adjust pH to 3 with HCl)
SMILES FC1=CC=C(C2=C(C3=CC=CC=C3)C(C(NC4=CC=CC=C4O)=O)=C(C(C)C)N2CC[C@@H](O)C[C@@H](O)CC(O)=O)C=C1.[Ca].[1/2]
Molecular Weight (g/mol) 594.69
Formula Weight 594.69
Percent Purity 95.92%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.