Learn More
2-Heptyne, 98+%, Thermo Scientific™
Product Code. p-7025835
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
Product Code. 11467094
Brand: Thermo Scientific Alfa Aesar B21528.04
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1119-65-9
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00039970
InChI Key AMSFEMSYKQQCHL-UHFFFAOYSA-N
Synonym 2-heptyne, 1-methyl-2-butylacetylene, butylmethylacetylene, n-butylmethylacetylene, acmc-1brzr, n-c4h9c.$.cch3, 2-heptyne,
PubChem CID 14245
IUPAC Name hept-2-yne
SMILES CCCCC#CC
Specifications
Density 0.753
Boiling Point 112°C to 113°C
Flash Point 11°C (52°F)
Odor Characteristic
Linear Formula CH3(CH2)3C≡CCH3
Refractive Index 1.422
Quantity 2 g
UN Number UN3295
Beilstein 1733563
Formula Weight 96.17
Percent Purity ≥98%
Chemical Name or Material 2-Heptyne