missing translation for 'onlineSavingsMsg'
Learn More

2-Heptanone, 99%

Product Code. p-7080585
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
This item is not returnable. View return policy

Product Code. 11469512

Brand: Thermo Scientific Alfa Aesar A10200.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-43-0 | C7H14O | 114.19 g/mol

2-Heptanone is used as a solvent for resins and lacquers. It is also used as a fragrance in cosmetics and a flavor for foods. It is involved as a reagent in the synthesis of rifamycin derivatives, which exhibit antibacterial activity. Further, it is employed in the preparation of thiazolidin-4-one derivatives as inhibitors of Toxoplasma gondii. In addition to this, it is considered as an alternative local anesthetic to lidocaine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Heptanone is used as a solvent for resins and lacquers. It is also used as a fragrance in cosmetics and a flavor for foods. It is involved as a reagent in the synthesis of rifamycin derivatives, which exhibit antibacterial activity. Further, it is employed in the preparation of thiazolidin-4-one derivatives as inhibitors of Toxoplasma gondii. In addition to this, it is considered as an alternative local anesthetic to lidocaine.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-43-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009513
InChI Key CATSNJVOTSVZJV-UHFFFAOYSA-N
Synonym 2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone
PubChem CID 8051
ChEBI CHEBI:5672
IUPAC Name heptan-2-one
SMILES CCCCCC(C)=O

Specifications

Melting Point -35°C
Density 0.82
Boiling Point 149°C to 150°C
Flash Point 47°C (116°F)
Odor Unpleasant
Refractive Index 1.409
Quantity 100 mL
UN Number UN1110
Beilstein 1699063
Merck Index 14,4663
Solubility Information Slightly miscible with water.
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material 2-Heptanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Heptanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 2-Heptanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.