missing translation for 'onlineSavingsMsg'
Learn More

2-Formylbenzenesulfonic acid sodium salt hydrate, tech. 90%, Thermo Scientific™

Product Code. 11488617
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488617 50 g 50g
11498617 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488617 Supplier Thermo Scientific Alfa Aesar Supplier No. A19736.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305808-14-4
Molecular Formula C7H5NaO4S
Molecular Weight (g/mol) 208.16
MDL Number MFCD00007478
InChI Key ADPUQRRLAAPXGT-UHFFFAOYSA-M
Synonym sodium 2-formylbenzenesulfonic acid hydrate
PubChem CID 87238610
IUPAC Name 2-formylbenzenesulfonic acid;sodium;hydrate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O

Specifications

Quantity 50 g
Sensitivity Air Sensitive
Formula Weight 208.17 (Anhydrous)
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Formylbenzenesulfonic acid sodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.