Learn More
2-Ethylthiophene, 99%
Product Code. 11471003
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471003 25 g 25g
11481003 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11471003 Supplier Thermo Scientific Alfa Aesar Supplier No. A13372.14
Request Bulk or Custom Format
CAS: 872-55-9 | C6H8S | 112.19 g/mol
It is used as a heterocyclic building block. Also a maillard product used to provide a pleasant aroma to meat and coffee.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a heterocyclic building block. Also a maillard product used to provide a pleasant aroma to meat and coffee.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 872-55-9
Molecular Formula C6H8S
Molecular Weight (g/mol) 112.19
MDL Number MFCD00005461
InChI Key JCCCMAAJYSNBPR-UHFFFAOYSA-N
Synonym thiophene, 2-ethyl, 2-ethyl-thiophene, ethylthiophene, thiophene, ethyl, pubchem5536, acmc-209qjc, 2-ethylthiophene, 2-ethylthiophene, analytical standard, met 2-cl-4-metpyrimidine-6-carboxylate
PubChem CID 13388
IUPAC Name 2-ethylthiophene
SMILES CCC1=CC=CS1
Specifications
Density 0.991
Boiling Point 133°C to 134°C
Flash Point 21°C (69°F)
Odor Stench
Refractive Index 1.512
Quantity 25 g
UN Number UN1993
Beilstein 105402
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 112.19
Percent Purity 99%
Chemical Name or Material 2-Ethylthiophene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Ethylthiophene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Ethylthiophene