Learn More
2-Dimethylaminobenzoic acid, 98%
Product Code. 11469823
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11469823
Brand: Thermo Scientific Alfa Aesar A12967.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 610-16-2 | C9H11NO2 | 165.192 g/mol
2-Dimethylaminobenzoic acid is used as a chemical and organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Dimethylaminobenzoic acid is used as a chemical and organic intermediate.
Solubility
Insoluble in water.
Notes
Store in tightly sealed containers in a cool place. Protect from moisture. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 610-16-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00016496
InChI Key DVVXXHVHGGWWPE-UHFFFAOYSA-N
Synonym 2-dimethylamino benzoic acid, n,n-dimethylanthranilic acid, benzoic acid, 2-dimethylamino, anthranilic acid, n,n-dimethyl, anthranilic acid,n-dimethyl, n,n-dimethylaminobenzoic acid, benzoic acid, dimethylamino, o-dimethylaminobenzoic acid, anthranilic acid, n,n-dimethyl-8ci, 2-dimethylamino-benzoic acid; hydrochloride
PubChem CID 69118
IUPAC Name 2-(dimethylamino)benzoic acid
SMILES CN(C)C1=CC=CC=C1C(=O)O
Specifications
Melting Point 70°C to 74°C
Odor Odorless
Quantity 1 g
Beilstein 2089466
Solubility Information Insoluble in water.
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material 2-Dimethylaminobenzoic acid