missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxyuridine, 99+%

Product Code. p-3757394 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10398120 250 mg Glass bottle 250mg
10115050 1 g Glass bottle 1g
10186633 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10398120 Supplier Thermo Scientific Chemicals Supplier No. 226612500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 951-78-0 | C9H12N2O5 | 228.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 951-78-0
Molecular Formula C9H12N2O5
Molecular Weight (g/mol) 228.20
MDL Number MFCD00006527
InChI Key MXHRCPNRJAMMIM-SHYZEUOFSA-N
Synonym 2'-deoxyuridine, deoxyuridine, uracil deoxyriboside, 2-deoxyuridine, 1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, deoxyribose uracil, uridine, 2'-deoxy, 2'-desoxyuridine, unii-w78i7ay22c, ccris 2832
PubChem CID 13712
ChEBI CHEBI:16450
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(=O)NC1=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 163.0°C to 167.0°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 2911
Solubility Information Solubility in water: 300g/L (20°C)
Formula Weight 228.2
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 2'-Deoxyuridine
Show More Show Less
compliance-icons
Product Identifier
  • 2'-Deoxyuridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.