missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyadenosine-5'-triphosphate disodium salt, 98%, Thermo Scientific™

Product Code. 15450337
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450337 100 mg 100mg
15440337 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450337 Supplier Thermo Scientific Alfa Aesar Supplier No. J64045.MC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2′-Deoxyadenosine-5′-triphosphate disodium salt is used as a substrate by a variety of polymerases including DNA polymerase(s) and reverse transcriptase(s).

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74299-50-6
Molecular Formula C10H14N5Na2O12P3
Molecular Weight (g/mol) 535.15
MDL Number MFCD00080335
InChI Key JEKDCIBJADJZSK-DZVPOJRENA-L
Synonym 2'-deoxyadenosine-5'-triphosphate disodium salt
PubChem CID 131674273
IUPAC Name disodium;[[[(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate
SMILES [Na+].[Na+].NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)O3)C2=NC=N1

Specifications

Color White
Quantity 100 mg
Solubility Information Soluble in water.
Formula Weight 535.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2'-Deoxyadenosine-5'-triphosphate disodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.