missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyadenosine-5'-Monophosphate Disodium Salt Hydrate, ≥98%, MP Biomedicals™

Product Code. 11402202 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402202 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11402202 Supplier MP Biomedicals Supplier No. 0215079501

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Soluble in water (50mg/mL)

TRUSTED_SUSTAINABILITY

Specifications

CAS 151151-31-4
Color White
Assay Percent Range ≥98%
Molecular Formula C10H12N5Na2O6P
MDL Number MFCD00065990
Quantity 1 g
Synonym 2'-deoxyadenosine-5'-monophosphate disodium salt, sodium 2r,3s,5r-5-6-amino-9h-purin-9-yl-3-hydroxytetrahydrofuran-2-yl methyl phosphate, damp hydrate, disodium 2'-deoxyadenosine 5'-phosphate, 2'-deoxyadenosine 5'-monophosphate disodium salt, 5'-adenylic acid, 2'-deoxy-, disodium salt, innoiuuoea, damp.2na, 2-deoxyadenosine-5-monophosphatedisodiumsalt, 2'-deoxyadenosine 5'-monophosphate sodium salt
InChI Key SNYHHRDGEJPLGT-OJSHLMAWSA-L
SMILES C1C(C(OC1N2C=NC3=C2N=CN=C3N)COP(=O)([O-])[O-])O.[Na+].[Na+]
IUPAC Name disodium;[(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methyl phosphate
Molecular Weight (g/mol) 375.188
PubChem CID 14598610
Formula Weight 375.2
Physical Form Powder
Chemical Name or Material 2'-Deoxyadenosine-5'-Monophosphate Disodium Salt Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.