missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopropyl-4-phenylquinoline-3-carbaldehyde, TRC

Product Code. 30353146
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353146 100 mg 100mg
30368381 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30353146 Supplier Toronto Research Chemicals Supplier No. TRCC989590100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2153433-59-9
Molecular Formula C19H15NO
Molecular Weight (g/mol) 273.33
Synonym 2-Cyclopropyl-4-phenyl-3-quinolinecarboxaldehyde (ACI)
IUPAC Name 2-cyclopropyl-4-phenylquinoline-3-carbaldehyde
SMILES O=CC1=C(C2=CC=CC=C2)C3=C(C=CC=C3)N=C1C4CC4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H15NO/c21-12-16-18(13-6-2-1-3-7-13)15-8-4-5-9-17(15)20-19(16)14-10-11-14/h1-9,12,14H,10-11H2
Formula Weight 273.12
Chemical Name or Material 2-Cyclopropyl-4-phenylquinoline-3-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.