missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentylcyclopentanone, 99%, Thermo Scientific™

Product Code. p-7045929
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11363196

Brand: Thermo Scientific Alfa Aesar L12503.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4884-24-6
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00051419
InChI Key CWZGKTMWPFTJCS-UHFFFAOYNA-N
Synonym 2-cyclopentylcyclopentanone, 1,1'-bicyclopentyl-2-one, bicyclopentyl-2-one, 1,1'-bi cyclopentan-2-one, bicyclopentylone, acmc-1aqra, 2-cyclopentyl cyclopentanone, cyclopentanone, 2-cyclopentyl
PubChem CID 21003
IUPAC Name 2-cyclopentylcyclopentan-1-one
SMILES O=C1CCCC1C1CCCC1

Specifications

Density 0.98
Boiling Point 230°C to 233°C
Flash Point 104°C (219°F)
Refractive Index 1.478
Quantity 5 g
Beilstein 1306869
Formula Weight 152.24
Percent Purity 99%
Chemical Name or Material 2-Cyclopentylcyclopentanone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.