missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)naphthalene, 98%

Product Code. p-7033187
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479360 250 mg 250mg
11489360 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479360 Supplier Thermo Scientific Alfa Aesar Supplier No. H50242.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 2506-41-4 | C11H9Cl | 176.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2506-41-4
Molecular Formula C11H9Cl
Molecular Weight (g/mol) 176.64
MDL Number MFCD00046488
InChI Key MPCHQYWZAVTABQ-UHFFFAOYSA-N
Synonym 2-chloromethyl naphthalene, naphthalene, 2-chloromethyl, 2-chloroethyl naphthalene, 2-chloromethyl-naphthalene, acmc-1ccgj, beta-chloromethylnapthalene, 3-chloro-methylnaphthalene, beta-chloromethylnaphthalene
PubChem CID 75632
IUPAC Name 2-(chloromethyl)naphthalene
SMILES ClCC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 47°C to 50°C
Quantity 250 mg
UN Number UN3261
Formula Weight 176.65
Percent Purity 98%
Chemical Name or Material 2-(Chloromethyl)naphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.