Learn More
2-Chloro-6-morpholinopyran-4-one (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-6-morpholinopyran-4-one |
| CAS | 119671-47-5 |
| Molecular Formula | C9H10ClNO3 |
| Quantity | 250 mg |
| Synonym | 2-Chloro-6-(4-morpholinyl)-4H-pyran-4-one, 2-Chloro-6-(morpholin-4-yl)-4H-pyran-4-one, 2-Chloro-6-(morpholin-4-yl)pyran-4-one |
| InChI Formula | InChI=1S/C9H10ClNO3/c10-8-5-7(12)6-9(14-8)11-1-3-13-4-2-11/h5-6H,1-4H2 |
| SMILES | ClC1=CC(=O)C=C(O1)N2CCOCC2 |
| IUPAC Name | 2-chloro-6-morpholin-4-ylpyran-4-one |
| Molecular Weight (g/mol) | 215.63 |
| Formula Weight | 215.0349 |