missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzonitrile, 98%, Thermo Scientific™

Product Code. 11420237
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420237 10 g 10g
11430237 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420237 Supplier Thermo Scientific Alfa Aesar Supplier No. B21662.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 668-45-1
Molecular Formula C7H3ClFN
Molecular Weight (g/mol) 155.556
MDL Number MFCD00001780
InChI Key XPTAYRHLHAFUOS-UHFFFAOYSA-N
Synonym 2-fluoro-6-chlorobenzonitrile, benzonitrile, 2-chloro-6-fluoro, 2-chloro-6-fluoro-benzonitrile, ncr bg ff, pubchem3423, ksc490q6t, acmc-1b036, timtec-bb sbb042501, 2-chloro-6-fluorobenzenecarbonitrile, 6-chloro-2-fluorobenzenecarbonitrile
PubChem CID 69587
IUPAC Name 2-chloro-6-fluorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)C#N)F

Specifications

Melting Point 56°C to 60°C
Quantity 10 g
Odor Characteristic
UN Number UN3439
Beilstein 1940332
Boiling Point 104°C to 105°C (11 mmHg)
Formula Weight 155.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.