missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5,6-dimethyl Nicotinic Acid, TRC

Product Code. 30354932
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30354932 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354932 Supplier Toronto Research Chemicals Supplier No. TRCC365425500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-5,6-dimethyl Nicotinic Acid
CAS 120003-75-0
Molecular Formula C8H8ClNO2
Quantity 500 mg
Synonym 2-Chloro-5,6-dimethyl-3-pyridinecarboxylic Acid
InChI Formula InChI=1S/C8H8ClNO2/c1-4-3-6(8(11)12)7(9)10-5(4)2/h3H,1-2H3,(H,11,12)
SMILES Cc1cc(C(=O)O)c(Cl)nc1C
IUPAC Name 2-chloro-5,6-dimethylpyridine-3-carboxylic acid
Molecular Weight (g/mol) 185.61
Formula Weight 185.0244
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.