missing translation for 'onlineSavingsMsg'
Learn More

2-[(Carboxymethyl)amino]propanoic Acid Hydrochloride, TRC

Product Code. 30366791
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366791 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366791 Supplier Toronto Research Chemicals Supplier No. TRCC183785250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(Carboxymethyl)amino]propanoic Acid Hydrochloride
CAS 33012-75-8
Molecular Formula C5H9NO4 • HCl
Quantity 250 mg
Synonym N-(Carboxymethyl)-L-alanine Hydrochloride, Strombine Hydrochloride
InChI Formula InChI=1S/C5H9NO4.ClH/c1-3(5(9)10)6-2-4(7)8;/h3,6H,2H2,1H3,(H,7,8)(H,9,10);1H/t3-;/m0./s1
SMILES [C@@H](NCC(O)=O)(C(O)=O)C.Cl
IUPAC Name (carboxymethyl)alanine hydrochloride
Molecular Weight (g/mol) 183.59
Formula Weight 183.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.