Learn More
2-Butyl-5-hydroxy-1H-imidazole, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Butyl-5-hydroxy-1H-imidazole |
| CAS | 1026802-97-0 |
| Molecular Formula | C7H12N2O |
| Quantity | 1 g |
| Synonym | 2-Butyl-1H-imidazol-5-ol |
| InChI Formula | InChI=1S/C7H12N2O/c1-2-3-4-6-8-5-7(10)9-6/h5,10H,2-4H2,1H3,(H,8,9) |
| SMILES | CCCCc1ncc(O)[nH]1 |
| IUPAC Name | 2-butyl-1H-imidazol-5-ol |
| Molecular Weight (g/mol) | 140.18 |
| Formula Weight | 140.095 |