missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenetole, TRC

Product Code. 30337194
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30337194 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337194 Supplier Toronto Research Chemicals Supplier No. TRCB686163500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 2-Bromophenetole
CAS 583-19-7
Molecular Formula C8H9BrO
Quantity 500 mg
InChI Formula InChI=1S/C8H9BrO/c1-2-10-8-6-4-3-5-7(8)9/h3-6H,2H2,1H3
SMILES CCOc1ccccc1Br
IUPAC Name 1-bromo-2-ethoxybenzene
Molecular Weight (g/mol) 201.06
Formula Weight 199.9837
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.