Learn More
2-Bromophenetole, TRC
Product Code. 30343128
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30343128 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30343128 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCB686163100MG
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material 2-Bromophenetole
CAS 583-19-7
Molecular Formula C8H9BrO
Quantity 100 mg
InChI Formula InChI=1S/C8H9BrO/c1-2-10-8-6-4-3-5-7(8)9/h3-6H,2H2,1H3
SMILES CCOc1ccccc1Br
IUPAC Name 1-bromo-2-ethoxybenzene
Molecular Weight (g/mol) 201.06
Formula Weight 199.9837