missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromoisophthaldehyde, TRC
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Specifications
Specifications
| Chemical Name or Material | 2-Bromoisophthaldehyde |
| CAS | 79839-49-9 |
| Molecular Formula | C8H5BrO2 |
| Quantity | 25 g |
| Synonym | 1-Bromo-2,6-diformylbenzene, 2-Bromo-1,3-benzenedicarboxaldehyde, 1-Bromo-2,6-diformylbenzene, 2-Bromobenzene-1,3-dialdehyde, 2-Bromobenzene-1,3-dicarboxaldehyde |
| InChI Formula | InChI=1S/C8H5BrO2/c9-8-6(4-10)2-1-3-7(8)5-11/h1-5H |
| SMILES | BrC1=C(C=O)C=CC=C1C=O |
| IUPAC Name | 2-bromoisophthalaldehyde |
| Molecular Weight (g/mol) | 213.03 |
| Formula Weight | 211.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction