Learn More
Alfa Aesar™ 2-Bromo-4,5-difluorobenzoic acid, 97%
Product Code. 15426888
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15426888 5 g 5g
15436888 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15426888 Supplier Alfa Aesar™ Supplier No. H60418.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 64695-84-7
Molecular Formula C7H3BrF2O2
Molecular Weight (g/mol) 237
MDL Number MFCD04973422
InChI Key DGCBGBZYTNTZJH-UHFFFAOYSA-N
PubChem CID 2782789
IUPAC Name 2-bromo-4,5-difluorobenzoic acid
SMILES C1=C(C(=CC(=C1F)F)Br)C(=O)O
Specifications
Melting Point 118°C to 122°C
Quantity 5 g
Synonym 2-bromo-4,5-difluorobenzoicacid, 2-bromo-4,5-difluoro-benzoic acid, benzoic acid, 2-bromo-4,5-difluoro, pubchem4991, acmc-209nnq, ksc495k3l, benzoic acid,2-bromo-4,5-difluoro, 2-bromo-4,5-difluorobenzoic acid, 2-bromanyl-4,5-bis fluoranyl benzoic acid
Formula Weight 237
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,5-difluorobenzoic acid