Learn More
2-Bromo-3-methoxy-6-nitropyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-3-methoxy-6-nitropyridine |
| CAS | 76066-07-4 |
| Molecular Formula | C6 H5 Br N2 O3 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C6H5BrN2O3/c1-12-4-2-3-5(9(10)11)8-6(4)7/h2-3H,1H3 |
| SMILES | COc1ccc(nc1Br)[N+](=O)[O-] |
| IUPAC Name | 2-bromo-3-methoxy-6-nitropyridine |
| Molecular Weight (g/mol) | 233.02 |
| Formula Weight | 231.9484 |