Learn More
2-Bromo-2'-(trifluoromethyl)acetophenone, 97%
Description
The sulfonyl urea thus obtained was used directly in the following condensation with 4-(trifluoromethyl)phenacyl bromide. Upon heating, benzenesulfinic acid ammonium salt might react with 2-(trifluoromethyl)phenacyl bromide to give β-keto sulfone. In Rap-Stoermer condensation reation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
The sulfonyl urea thus obtained was used directly in the following condensation with 4-(trifluoromethyl)phenacyl bromide. Upon heating, benzenesulfinic acid ammonium salt might react with 2-(trifluoromethyl)phenacyl bromide to give β-keto sulfone. In Rap-Stoermer condensation reation.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 24°C to 32°C |
| Quantity | 1 g |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 267.04 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Bromo-2'-(trifluoromethyl)acetophenone |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.