Learn More
2-Bromo-1,4-naphthoquinone, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-1,4-naphthoquinone |
| CAS | 2065-37-4 |
| Molecular Formula | C10H5BrO2 |
| Quantity | 1 g |
| Synonym | 2-Bromo-1,4-naphthalenedione, 2-Bromo-p-naphthoquinone, 2-Bromonaphthoquinone, 3-Bromo-1,4-naphthoquinone, NSC 401097 |
| InChI Formula | InChI=1S/C10H5BrO2/c11-8-5-9(12)6-3-1-2-4-7(6)10(8)13/h1-5H |
| SMILES | BrC1=CC(=O)c2ccccc2C1=O |
| IUPAC Name | 2-bromonaphthalene-1,4-dione |
| Molecular Weight (g/mol) | 237.05 |