missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(3,4-dimethoxyphenyl)ethanone, TRC

Product Code. 30347356
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347356 1 g 1g
30333542 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30347356 Supplier Toronto Research Chemicals Supplier No. TRCB6840751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromo-1-(3,4-dimethoxyphenyl)ethanone
CAS 1835-02-5
Molecular Formula C10H11BrO3
Quantity 1 g
Synonym 2-Bromo-1-(3,4-dimethoxyphenyl)ethanone, 2-Bromo-3',4'-dimethoxyacetophenone, 2-Bromoacetoveratrone, 3,4-Dimethoxyphenacyl Bromide, 3',4'-Dimethoxy-2-bromoacetophenone, NSC 112833
InChI Formula InChI=1S/C10H11BrO3/c1-13-9-4-3-7(8(12)6-11)5-10(9)14-2/h3-5H,6H2,1-2H3
SMILES COc1ccc(cc1OC)C(=O)CBr
IUPAC Name 2-bromo-1-(3,4-dimethoxyphenyl)ethanone
Molecular Weight (g/mol) 259.1
Formula Weight 257.9892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.