Learn More
2-(Benzyloxy)-1-ethanamine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(Benzyloxy)-1-ethanamine |
| CAS | 38336-04-8 |
| Molecular Formula | C9 H13 N O |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C9H13NO/c10-6-7-11-8-9-4-2-1-3-5-9/h1-5H,6-8,10H2 |
| SMILES | NCCOCc1ccccc1 |
| IUPAC Name | 2-benzyloxyethanamine |
| Molecular Weight (g/mol) | 151.21 |
| Formula Weight | 151.0997 |