Learn More
[2-(Aminocarbonyl)phenoxy]acetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | [2-(Aminocarbonyl)phenoxy]acetic Acid |
| CAS | 25395-22-6 |
| Molecular Formula | C9H9NO4 |
| Quantity | 2.5 g |
| Synonym | 2-[2-(Aminocarbonyl)phenoxy]acetic Acid, (2-Carbamoylphenoxy)acetic Acid, (o-Carbamoylphenoxy)acetic Acid, 2-(2-Carbamoylphenoxy)acetic Acid, 2-(2-Carbamoylphenoxy)aceticacid, 2-(Carboxymethoxy)benzamide, NSC 65589Salicylamide O-Acetic Acid, [2-(Aminocarbonyl)phenoxy]acetic Acid, o-Carboxyamidophenoxyacetic Acid |
| InChI Formula | InChI=1S/C9H9NO4/c10-9(13)6-3-1-2-4-7(6)14-5-8(11)12/h1-4H,5H2,(H2,10,13)(H,11,12) |
| SMILES | NC(=O)c1ccccc1OCC(=O)O |
| IUPAC Name | 2-(2-carbamoylphenoxy)acetic acid |
| Molecular Weight (g/mol) | 195.17 |
| Formula Weight | 195.0532 |