missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzeneboronic acid pinacol ester hydrochloride, 96%, Thermo Scientific™

Product Code. 15470466
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470466 250 mg 250mg
15460466 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470466 Supplier Thermo Scientific Alfa Aesar Supplier No. H53194.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393877-09-3
Molecular Formula C12H19BClNO2
Molecular Weight (g/mol) 255.549
MDL Number MFCD02179449
InChI Key XWILEMYKORLIPW-UHFFFAOYSA-N
Synonym 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline hydrochloride, 2-aminophenylboronic acid, pinacol ester, hcl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline hydrochloride, 2-aminobenzeneboronic acid pinacol ester hci, 2-aminophenyl boronic acid, pinacol ester hydrochloride, 2-aminophenyl boronic acid,pinacol ester hydrochloride, 2-aminobenzeneboronic acid pinacol ester hydrochloride, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline-hydrogen chloride 1/1, benzenamine, 2-4,4,5,5-tetremethyl-1,3,2-dioxaborolan-2-yl-, hcl salt
PubChem CID 44118691
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline;hydrochloride
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2N.Cl

Specifications

Melting Point 161°C to 163°C
Quantity 250 mg
Formula Weight 255.55
Percent Purity 96%
Chemical Name or Material 2-Aminobenzeneboronic acid pinacol ester hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.