Learn More
2-Amino-5-fluorobenzothiazole, 98%
Product Code. p-7033844
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15457907
Brand: Thermo Scientific Alfa Aesar H51022.MD
Request Bulk or Custom Format
CAS: 20358-07-0 | C7H5FN2S | 168.189 g/mol
2-Amino-5-fluorobenzothiazole is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-5-fluorobenzothiazole is used as organic chemical synthesis intermediate.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20358-07-0
Molecular Formula C7H5FN2S
Molecular Weight (g/mol) 168.189
MDL Number MFCD05664560
InChI Key YHBIGBYIUMCLJS-UHFFFAOYSA-N
PubChem CID 821203
IUPAC Name 5-fluoro-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1F)N=C(S2)N
Specifications
Quantity 250 mg
Synonym 2-amino-5-fluorobenzothiazole, 5-fluorobenzo d thiazol-2-amine, 2-benzothiazolamine, 5-fluoro, 5-fluoro-2-benzothiazolamine, 2-benzothiazolamine,5-fluoro, 5-fluorobenzothiazol-2-ylamine, pubchem21850, acmc-20a67y, 2-amino-5-fluorobenzothiozole, 5-fluoro-2-aminobenzothiazole
Solubility Information Soluble in methanol.
Formula Weight 168.19
Percent Purity 98%
Chemical Name or Material 2-Amino-5-fluorobenzothiazole