Learn More
2-Amino-4,5-dimethoxybenzonitrile, 98%, Thermo Scientific™
Product Code. 11337486
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337486 25 g 25g
11327486 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11337486 Supplier Thermo Scientific Alfa Aesar Supplier No. L09545.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26961-27-3
Molecular Formula C9H10N2O2
Molecular Weight (g/mol) 178.19
MDL Number MFCD00058682
InChI Key BJAYMNUBIULRMF-UHFFFAOYSA-N
Synonym 6-aminoveratronitrile, 2-cyano-4,5-dimethoxyaniline, 4-amino-5-cyanoveratrole, 6-amino-3,4-dimethoxybenzonitrile, benzonitrile, 2-amino-4,5-dimethoxy, 2-amino-4,5-dimethoxy-benzonitrile, 2-amino-4,5-dimethoxybenzenecarbonitrile, pubchem4632, acmc-209wvj
PubChem CID 735967
IUPAC Name 2-amino-4,5-dimethoxybenzonitrile
SMILES COC1=C(OC)C=C(C#N)C(N)=C1
Specifications
Melting Point 97°C to 101°C
Quantity 25 g
UN Number UN3439
Beilstein 2372149
Formula Weight 178.19
Percent Purity 98%
Chemical Name or Material 2-Amino-4,5-dimethoxybenzonitrile