missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylpyrazine, 99%

Product Code. p-7022929
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11488446

Brand: Thermo Scientific Alfa Aesar A15917.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22047-25-2 | C6H6N2O | 122.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22047-25-2
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.127
MDL Number MFCD00006134
InChI Key DBZAKQWXICEWNW-UHFFFAOYSA-N
Synonym acetylpyrazine, 2-acetylpyrazine, ethanone, 1-pyrazinyl, 1-pyrazin-2-yl ethan-1-one, methyl pyrazinyl ketone, 2-acetyl pyrazine, ketone, methyl pyrazinyl, 1-pyrazinylethanone, 1-pyrazin-2-yl-ethanone, 1-pyrazin-2-yl ethanone
PubChem CID 30914
IUPAC Name 1-pyrazin-2-ylethanone
SMILES CC(=O)C1=NC=CN=C1

Specifications

Melting Point 75°C to 78°C
Boiling Point 78°C to 79°C (8 mmHg)
Quantity 1 g
Beilstein 109630
Formula Weight 122.13
Percent Purity 99%
Chemical Name or Material 2-Acetylpyrazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.