missing translation for 'onlineSavingsMsg'
Learn More

2'-Acetoxyacetophenone, 98%

Product Code. p-7025897
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11427254

Brand: Thermo Scientific Alfa Aesar B21624.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7250-94-4 | C10H10O3 | 178.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7250-94-4
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00017223
InChI Key WWEDAIJJBDFOOK-UHFFFAOYSA-N
Synonym 2'-acetoxyacetophenone, 2-acetylphenyl acetate, ethanone, 1-2-acetyloxy phenyl, o-acetoxyacetophenone, 2'-acetoxy acetophenone, maybridge1_002653, acmc-209on8, acetic acid 2-acetyl-phenyl ester, acetophenone, 2-hydroxy-, acetate 8ci
PubChem CID 16716
IUPAC Name (2-acetylphenyl) acetate
SMILES CC(=O)OC1=CC=CC=C1C(C)=O

Specifications

Melting Point 88°C to 91°C
Boiling Point 118°C (19 mmHg)
Quantity 5 g
Beilstein 1948938
Formula Weight 178.19
Percent Purity 98%
Chemical Name or Material 2'-Acetoxyacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.