missing translation for 'onlineSavingsMsg'
Learn More

(2-Acetamido-5-chlorophenyl)(phenyl)carbamic Acid tert-Butyl Ester, TRC

Product Code. 30315542
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315542 25 mg 25mg
30324760 50 mg 50mg
30313220 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30315542 Supplier Toronto Research Chemicals Supplier No. TRCA15010525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2-Acetamido-5-chlorophenyl)(phenyl)carbamic Acid tert-Butyl Ester
Analyte or Component Names (2-Acetamido-5-chlorophenyl)(phenyl)carbamic Acid tert-Butyl Ester
Shipping Condition Room Temperature
Molecular Formula C19H21ClN2O3
Synonym (2-Acetamido-5-chlorophenyl)(phenyl)carbamic Acid tert-Butyl Ester
InChI Formula InChI=1S/C19H21ClN2O3/c1-13(23)21-16-11-10-14(20)12-17(16)22(15-8-6-5-7-9-15)18(24)25-19(2,3)4/h5-12H,1-4H3,(H,21,23)
Solution Type Neat
SMILES ClC1=CC=C(NC(C)=O)C(N(C2=CC=CC=C2)C(OC(C)(C)C)=O)=C1
IUPAC Name tert-butyl (2-acetamido-5-chlorophenyl)(phenyl)carbamate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 360.84
Quantity 25 mg
Formula Weight 360.12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.